Assay Details
| Gene Symbol: alox5ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032320.1 | NP_001027491.1 | 3 - 4 | 270 | 65 | |
| GenBank mRNA | BC096503.1 | - | 3 - 4 | 270 | 65 | |
More Information
Set Membership: |
|

alox5ap
Genomic Map