Assay Details
| Gene Symbol: xkr6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033943.1 | NP_001029115.1 | 2 - 3 | 898 | 90 | |
| XM_018089355.1 | XP_017944844.1 | 4 - 5 | 1885 | 90 | ||
| GenBank mRNA | AY699709.1 | - | 2 - 3 | 898 | 90 | |
More Information
Set Membership: |
|

xkr6
Genomic Map