Assay Details
| Gene Symbol: igfbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033946.1 | NP_001029118.1 | 2 - 3 | 610 | 83 | |
| GenBank mRNA | BC099978.1 | - | 2 - 3 | 610 | 83 | |
More Information
Set Membership: |
|

igfbp1
Genomic Map