Assay Details
| Gene Symbol: dusp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005450.2 | NP_001005450.2 | 1 - 2 | 595 | 64 | |
| GenBank mRNA | BC074564.1 | - | 1 - 2 | 472 | 64 | |
| CR760962.2 | - | 1 - 2 | 595 | 64 | ||
More Information
Set Membership: |
|

dusp1
Genomic Map