Assay Details
| Gene Symbol: coq6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006828.2 | NP_001006829.1 | 1 - 2 | 175 | 72 | |
| GenBank mRNA | BC076897.1 | - | 2 - 3 | 191 | 72 | |
| CR761032.2 | - | 1 - 2 | 175 | 72 | ||
More Information
Set Membership: |
|

coq6
Genomic Map