Assay Details
| Gene Symbol: anxa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008047.2 | NP_001008048.1 | 10 - 11 | 875 | 79 | |
| GenBank mRNA | BC080933.1 | - | 10 - 11 | 947 | 79 | |
| CR760221.2 | - | 10 - 11 | 875 | 79 | ||
More Information
Set Membership: |
|

anxa9
Genomic Map