Assay Details
| Gene Symbol: traf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008161.2 | NP_001008162.2 | 2 - 3 | 534 | 66 | |
| GenBank mRNA | BC082342.1 | - | 2 - 3 | 536 | 66 | |
| CR855449.2 | - | 2 - 3 | 534 | 66 | ||
More Information
Set Membership: |
|

traf6
Genomic Map