Assay Details
| Gene Symbol: pcmtd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011087.2 | NP_001011087.2 | 4 - 5 | 662 | 50 | |
| GenBank mRNA | BC084479.1 | - | 4 - 5 | 663 | 50 | |
| CR761105.1 | - | 4 - 5 | 662 | 50 | ||
More Information
Set Membership: |
|

pcmtd2
Genomic Map