Assay Details
| Gene Symbol: cpeb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015925.2 | NP_001015925.1 | 8 - 9 | 1764 | 63 | |
| XM_012966552.2 | XP_012822006.1 | 7 - 8 | 1974 | 63 | ||
| XM_018095384.1 | XP_017950873.1 | 9 - 10 | 1875 | 63 | ||
| XM_018095385.1 | XP_017950874.1 | 9 - 10 | 1643 | 63 | ||
| XM_018095386.1 | XP_017950875.1 | 7 - 8 | 1975 | 63 | ||
| GenBank mRNA | CR926390.2 | - | 8 - 9 | 1764 | 63 | |
More Information
Set Membership: |
|

cpeb3
Genomic Map