Assay Details
| Gene Symbol: pycr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015942.2 | NP_001015942.1 | 3 - 4 | 569 | 73 | |
| GenBank mRNA | CR848490.2 | - | 3 - 4 | 569 | 73 | |
| CR942403.2 | - | 3 - 4 | 278 | 73 | ||
More Information
Set Membership: |
|

pycr1
Genomic Map