Assay Details
| Gene Symbol: brk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015994.2 | NP_001015994.1 | 1 - 2 | 139 | 91 | |
| XM_018093034.1 | XP_017948523.1 | 2 - 3 | 224 | 91 | ||
| GenBank mRNA | BC167873.1 | - | 1 - 2 | 172 | 91 | |
| CR848305.2 | - | 1 - 2 | 139 | 91 | ||
More Information
Set Membership: |
|

brk1
Genomic Map