Assay Details
| Gene Symbol: crx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016021.2 | NP_001016021.1 | 2 - 3 | 372 | 81 | |
| XM_012968359.1 | XP_012823813.1 | 4 - 5 | 666 | 81 | ||
| GenBank mRNA | BC085002.1 | - | 2 - 3 | 483 | 81 | |
| BC161339.1 | - | 2 - 3 | 459 | 81 | ||
| BC170967.1 | - | 2 - 3 | 271 | 81 | ||
| BC170992.1 | - | 2 - 3 | 271 | 81 | ||
| CR848167.2 | - | 2 - 3 | 372 | 81 | ||
More Information
Set Membership: |
|

crx
Genomic Map