Assay Details
| Gene Symbol: ccndx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016108.2 | NP_001016108.1 | 4 - 5 | 763 | 50 | |
| GenBank mRNA | BC158907.1 | - | 4 - 5 | 766 | 50 | |
| CR762119.2 | - | 4 - 5 | 763 | 50 | ||
| DQ508541.1 | - | 4 - 5 | 763 | 50 | ||
More Information
Set Membership: |
|

ccndx
Genomic Map