Assay Details
| Gene Symbol: metap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016115.2 | NP_001016115.1 | 4 - 5 | 446 | 72 | |
| GenBank mRNA | BC161036.1 | - | 4 - 5 | 442 | 72 | |
| CR762101.2 | - | 4 - 5 | 446 | 72 | ||
More Information
Set Membership: |
|

metap2
Genomic Map