Assay Details
| Gene Symbol: kif11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016116.3 | NP_001016116.2 | 21 - 22 | 3141 | 59 | |
| XM_012966561.1 | XP_012822015.1 | 22 - 23 | 3301 | 59 | ||
| GenBank mRNA | BC166147.1 | - | 21 - 22 | 3072 | 59 | |
| CR762094.2 | - | 15 - 16 | 1887 | 59 | ||
| CX486424.1 | - | 2 - 3 | 541 | 59 | ||
| EF524557.1 | - | 20 - 21 | 2960 | 59 | ||
More Information
Set Membership: |
|

kif11
Genomic Map