Assay Details
| Gene Symbol: mrpl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016126.2 | NP_001016126.1 | 1 - 2 | 55 | 61 | |
| GenBank mRNA | BC098087.1 | - | 1 - 2 | 30 | 61 | |
| BC158989.1 | - | 1 - 2 | 33 | 61 | ||
| BC170718.1 | - | 1 - 2 | 55 | 61 | ||
| BC170720.1 | - | 1 - 2 | 55 | 61 | ||
| CR762063.2 | - | 1 - 2 | 55 | 61 | ||
More Information
Set Membership: |
|

mrpl1
Genomic Map