Assay Details
| Gene Symbol: tube1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016136.2 | NP_001016136.1 | 5 - 6 | 349 | 78 | |
| XM_012962819.2 | XP_012818273.1 | 5 - 6 | 559 | 78 | ||
| GenBank mRNA | BC121864.1 | - | 5 - 6 | 446 | 78 | |
| CR762030.2 | - | 5 - 6 | 349 | 78 | ||
More Information
Set Membership: |
|

tube1
Genomic Map