Assay Details
Gene Symbol: fam57a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016139.3 | NP_001016139.2 | 2 - 3 | 480 | 91 | |
GenBank mRNA | CR762024.2 | - | 2 - 3 | 210 | 91 | |
CU075575.1 | - | 2 - 3 | 480 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
