Assay Details
Gene Symbol: fam57a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016139.3 | NP_001016139.2 | 3 - 4 | 691 | 99 | |
GenBank mRNA | BX771253.1 | - | 2 - 3 | 651 | 99 | |
CR762024.2 | - | 3 - 4 | 421 | 99 | ||
CU075575.1 | - | 3 - 4 | 691 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
