Assay Details
Gene Symbol: fam57a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016139.3 | NP_001016139.2 | 4 - 5 | 787 | 67 | |
GenBank mRNA | BX771253.1 | - | 1 - 2 | 555 | 67 | |
CR762024.2 | - | 4 - 5 | 517 | 67 | ||
CU075575.1 | - | 4 - 5 | 787 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
