Assay Details
| Gene Symbol: sh3bgrl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016155.2 | NP_001016155.1 | 2 - 3 | 509 | 80 | |
| GenBank mRNA | BC077071.1 | - | 2 - 3 | 399 | 80 | |
| BC167267.1 | - | 2 - 3 | 382 | 80 | ||
| BC170711.1 | - | 2 - 3 | 442 | 80 | ||
| BC170713.1 | - | 2 - 3 | 442 | 80 | ||
| CR761949.2 | - | 2 - 3 | 509 | 80 | ||
More Information
Set Membership: |
|

sh3bgrl2
Genomic Map