Assay Details
| Gene Symbol: mrm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016220.2 | NP_001016220.1 | 3 - 4 | 788 | 79 | |
| GenBank mRNA | BC135861.1 | - | 3 - 4 | 727 | 79 | |
| CR926308.2 | - | 3 - 4 | 788 | 79 | ||
More Information
Set Membership: |
|

mrm3
Genomic Map