Assay Details
| Gene Symbol: kat8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016234.3 | NP_001016234.2 | 5 - 6 | 766 | 78 | |
| XM_018097101.1 | XP_017952590.1 | 5 - 6 | 1030 | 78 | ||
| GenBank mRNA | BC171095.1 | - | 5 - 6 | 628 | 78 | |
| BC171099.1 | - | 5 - 6 | 628 | 78 | ||
| CR926273.2 | - | 5 - 6 | 657 | 78 | ||
| CX846881.1 | - | 5 - 6 | 749 | 78 | ||
More Information
Set Membership: |
|

kat8
Genomic Map