Assay Details
| Gene Symbol: tor1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016237.2 | NP_001016237.1 | 2 - 3 | 526 | 76 | |
| GenBank mRNA | BC161275.1 | - | 2 - 3 | 461 | 76 | |
| BC170808.1 | - | 2 - 3 | 511 | 76 | ||
| BC171143.1 | - | 2 - 3 | 511 | 76 | ||
| CR926267.2 | - | 2 - 3 | 526 | 76 | ||
More Information
Set Membership: |
|

tor1b
Genomic Map