Assay Details
| Gene Symbol: sfxn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016244.2 | NP_001016244.1 | 10 - 11 | 959 | 68 | |
| GenBank mRNA | BC168602.1 | - | 10 - 11 | 926 | 68 | |
| BC170588.1 | - | 10 - 11 | 915 | 68 | ||
| BC170943.1 | - | 10 - 11 | 915 | 68 | ||
| CR761782.2 | - | 10 - 11 | 959 | 68 | ||
More Information
Set Membership: |
|

sfxn1
Genomic Map