Assay Details
| Gene Symbol: cby1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016253.2 | NP_001016253.1 | 5 - 6 | 486 | 65 | |
| XM_012961077.2 | XP_012816531.1 | 4 - 5 | 458 | 65 | ||
| XM_012961078.2 | XP_012816532.1 | 5 - 6 | 556 | 65 | ||
| GenBank mRNA | BC167320.1 | - | 5 - 6 | 547 | 65 | |
| BC170579.1 | - | 5 - 6 | 468 | 65 | ||
| BC170581.1 | - | 5 - 6 | 468 | 65 | ||
| CR761748.2 | - | 5 - 6 | 486 | 65 | ||
More Information
Set Membership: |
|

cby1
Genomic Map