Assay Details
| Gene Symbol: smim19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016254.3 | NP_001016254.2 | 3 - 4 | 352 | 69 | |
| NM_001244887.1 | NP_001231816.1 | 3 - 4 | 343 | 69 | ||
| XM_012971763.2 | XP_012827217.1 | 3 - 4 | 411 | 69 | ||
| GenBank mRNA | BC135856.1 | - | 3 - 4 | 285 | 69 | |
| CN120506.1 | - | 3 - 4 | 352 | 69 | ||
| CR421163.1 | - | 3 - 4 | 343 | 69 | ||
| CR761744.2 | - | 2 - 3 | 272 | 69 | ||
| CX373428.2 | - | 3 - 4 | 285 | 69 | ||
| CX514979.2 | - | 3 - 4 | 327 | 69 | ||
More Information
Set Membership: |
|

smim19
Genomic Map