Assay Details
| Gene Symbol: ccnh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016256.2 | NP_001016256.1 | 1 - 2 | 118 | 80 | |
| GenBank mRNA | BC118698.1 | - | 1 - 2 | 148 | 80 | |
| CR761723.2 | - | 1 - 2 | 118 | 80 | ||
More Information
Set Membership: |
|

ccnh
Genomic Map