Assay Details
| Gene Symbol: ier3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016260.2 | NP_001016260.1 | 106 | 70 | ||
| GenBank mRNA | CR761708.2 | - | 106 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ier3ip1| Gene Symbol: ier3ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016260.2 | NP_001016260.1 | 106 | 70 | ||
| GenBank mRNA | CR761708.2 | - | 106 | 70 | ||
Set Membership: |
|