Assay Details
| Gene Symbol: pym1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016274.2 | NP_001016274.1 | 2 - 3 | 150 | 96 | |
| GenBank mRNA | BC161580.1 | - | 136 | 96 | ||
| BC170597.1 | - | 2 - 3 | 150 | 96 | ||
| BC170601.1 | - | 2 - 3 | 150 | 96 | ||
| CR761629.2 | - | 2 - 3 | 150 | 96 | ||
More Information
Set Membership: |
|

pym1
Genomic Map