Assay Details
| Gene Symbol: kti12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016291.2 | NP_001016291.1 | 6 - 7 | 430 | 100 | |
| XM_012966570.1 | XP_012822024.1 | 6 - 7 | 624 | 100 | ||
| XM_018092083.1 | XP_017947572.1 | 6 - 7 | 463 | 100 | ||
| GenBank mRNA | BC170877.1 | - | 5 - 6 | 410 | 100 | |
| BC170879.1 | - | 5 - 6 | 410 | 100 | ||
| CR761570.2 | - | 6 - 7 | 430 | 100 | ||
More Information
Set Membership: |
|

kti12
Genomic Map