Assay Details
| Gene Symbol: dpm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016297.2 | NP_001016297.1 | 3 - 4 | 277 | 65 | |
| GenBank mRNA | BC157179.1 | - | 3 - 4 | 275 | 65 | |
| CR761535.2 | - | 3 - 4 | 277 | 65 | ||
More Information
Set Membership: |
|

dpm2
Genomic Map