Assay Details
| Gene Symbol: mrps16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016347.2 | NP_001016347.1 | 2 - 3 | 389 | 82 | |
| GenBank mRNA | BC158905.1 | - | 2 - 3 | 373 | 82 | |
| BC167288.1 | - | 2 - 3 | 373 | 82 | ||
| BC171007.1 | - | 2 - 3 | 340 | 82 | ||
| CR761358.2 | - | 2 - 3 | 389 | 82 | ||
More Information
Set Membership: |
|

mrps16
Genomic Map