Assay Details
| Gene Symbol: pabpn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016352.2 | NP_001016352.1 | 5 - 6 | 556 | 76 | |
| XM_012961085.2 | XP_012816539.1 | 6 - 7 | 875 | 76 | ||
| XM_012961086.2 | XP_012816540.1 | 6 - 7 | 642 | 76 | ||
| XM_012961087.2 | XP_012816541.1 | 6 - 7 | 661 | 76 | ||
| GenBank mRNA | BC121546.1 | - | 6 - 7 | 767 | 76 | |
| CR761333.2 | - | 5 - 6 | 556 | 76 | ||
More Information
Set Membership: |
|

pabpn1l
Genomic Map