Assay Details
| Gene Symbol: comt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016357.2 | NP_001016357.1 | 2 - 3 | 499 | 71 | |
| GenBank mRNA | CR761306.2 | - | 2 - 3 | 499 | 71 | |
More Information
Set Membership: |
|

comt
Genomic Map