Assay Details
| Gene Symbol: men1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016361.2 | NP_001016361.1 | 3 - 4 | 1226 | 72 | |
| GenBank mRNA | BC123035.1 | - | 3 - 4 | 1329 | 72 | |
| CR761297.2 | - | 3 - 4 | 1226 | 72 | ||
More Information
Set Membership: |
|

men1
Genomic Map