Assay Details
| Gene Symbol: atp5f1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016407.2 | NP_001016407.1 | 6 - 7 | 698 | 94 | |
| XM_012959013.1 | XP_012814467.1 | 6 - 7 | 781 | 94 | ||
| XM_012959014.1 | XP_012814468.1 | 6 - 7 | 781 | 94 | ||
| GenBank mRNA | CR761126.2 | - | 6 - 7 | 698 | 94 | |
More Information
Set Membership: |
|

atp5f1c
Genomic Map