Assay Details
| Gene Symbol: atp5f1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016407.2 | NP_001016407.1 | 8 - 9 | 958 | 103 | |
| GenBank mRNA | CR761126.2 | - | 8 - 9 | 958 | 103 | |
More Information
Set Membership: |
|

atp5f1c
Genomic Map