Assay Details
Gene Symbol: cd63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016413.2 | NP_001016413.1 | 7 - 8 | 762 | 60 | |
XM_018091249.1 | XP_017946738.1 | 7 - 8 | 971 | 60 | ||
GenBank mRNA | BC061271.1 | - | 7 - 8 | 695 | 60 | |
BC080508.1 | - | 7 - 8 | 742 | 60 | ||
BC135543.1 | - | 7 - 8 | 717 | 60 | ||
CR761109.2 | - | 7 - 8 | 762 | 60 | ||
CU464109.1 | - | 6 - 7 | 674 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
