Assay Details
| Gene Symbol: atp5me | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016415.2 | NP_001016415.1 | 3 - 4 | 301 | 85 | |
| GenBank mRNA | BC167416.1 | - | 3 - 4 | 199 | 85 | |
| CR761104.2 | - | 3 - 4 | 301 | 85 | ||
More Information
Set Membership: |
|

atp5me
Genomic Map