Assay Details
| Gene Symbol: efna5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016422.2 | NP_001016422.1 | 1 - 2 | 149 | 82 | |
| XM_012962281.2 | XP_012817735.1 | 1 - 2 | 2431 | 82 | ||
| GenBank mRNA | BC170814.1 | - | 1 - 2 | 148 | 82 | |
| BC170816.1 | - | 1 - 2 | 148 | 82 | ||
| CR761072.2 | - | 1 - 2 | 149 | 82 | ||
More Information
Set Membership: |
|

efna5
Genomic Map