Assay Details
| Gene Symbol: ca12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016431.2 | NP_001016431.1 | 9 - 10 | 1056 | 82 | |
| GenBank mRNA | BC135174.2 | - | 9 - 10 | 989 | 82 | |
| CR761051.2 | - | 9 - 10 | 1056 | 82 | ||
More Information
Set Membership: |
|

ca12
Genomic Map