Assay Details
| Gene Symbol: mif4gd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016440.2 | NP_001016440.1 | 3 - 4 | 262 | 77 | |
| XM_012971766.2 | XP_012827220.1 | 3 - 4 | 335 | 77 | ||
| XM_012971767.2 | XP_012827221.1 | 3 - 4 | 398 | 77 | ||
| XM_012971768.2 | XP_012827222.1 | 3 - 4 | 331 | 77 | ||
| XM_012971769.2 | XP_012827223.1 | 3 - 4 | 395 | 77 | ||
| XM_018097740.1 | XP_017953229.1 | 3 - 4 | 334 | 77 | ||
| GenBank mRNA | CR761027.2 | - | 3 - 4 | 262 | 77 | |
More Information
Set Membership: |
|

mif4gd
Genomic Map