Assay Details
| Gene Symbol: mif4gd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016440.2 | NP_001016440.1 | 4 - 5 | 421 | 100 | |
| XM_012971766.2 | XP_012827220.1 | 4 - 5 | 494 | 100 | ||
| XM_012971767.2 | XP_012827221.1 | 4 - 5 | 557 | 100 | ||
| XM_012971768.2 | XP_012827222.1 | 4 - 5 | 490 | 100 | ||
| XM_012971769.2 | XP_012827223.1 | 4 - 5 | 554 | 100 | ||
| GenBank mRNA | CR761027.2 | - | 4 - 5 | 421 | 100 | |
More Information
Set Membership: |
|

mif4gd
Genomic Map