Assay Details
| Gene Symbol: mif4gd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016440.2 | NP_001016440.1 | 5 - 6 | 513 | 67 | |
| XM_012971768.2 | XP_012827222.1 | 5 - 6 | 582 | 67 | ||
| XM_012971769.2 | XP_012827223.1 | 5 - 6 | 646 | 67 | ||
| GenBank mRNA | CR761027.2 | - | 5 - 6 | 513 | 67 | |
More Information
Set Membership: |
|

mif4gd
Genomic Map