Assay Details
| Gene Symbol: golga7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016450.3 | NP_001016450.1 | 3 - 4 | 388 | 72 | |
| XM_012959019.2 | XP_012814473.1 | 4 - 5 | 516 | 72 | ||
| XM_012959020.2 | XP_012814474.1 | 4 - 5 | 517 | 72 | ||
| GenBank mRNA | BC170747.1 | - | 3 - 4 | 387 | 72 | |
| BC170779.1 | - | 3 - 4 | 387 | 72 | ||
| CR760994.2 | - | 3 - 4 | 390 | 72 | ||
More Information
Set Membership: |
|

golga7
Genomic Map