Assay Details
Gene Symbol: arv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016452.2 | NP_001016452.1 | 4 - 5 | 478 | 69 | |
XM_012963009.2 | XP_012818463.1 | 4 - 5 | 572 | 69 | ||
GenBank mRNA | BC170566.1 | - | 3 - 4 | 422 | 69 | |
BC170568.1 | - | 3 - 4 | 422 | 69 | ||
CR760975.2 | - | 4 - 5 | 478 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
