Assay Details
| Gene Symbol: brcc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016457.2 | NP_001016457.1 | 7 - 8 | 653 | 70 | |
| GenBank mRNA | BC096514.1 | - | 7 - 8 | 645 | 70 | |
| CR760965.2 | - | 7 - 8 | 653 | 70 | ||
More Information
Set Membership: |
|

brcc3
Genomic Map