Assay Details
| Gene Symbol: stk17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016460.2 | NP_001016460.1 | 5 - 6 | 825 | 111 | |
| GenBank mRNA | BC171253.1 | - | 5 - 6 | 823 | 111 | |
| BC171255.1 | - | 5 - 6 | 823 | 111 | ||
| CR760956.2 | - | 5 - 6 | 825 | 111 | ||
More Information
Set Membership: |
|

stk17a
Genomic Map