Assay Details
| Gene Symbol: elmod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016465.2 | NP_001016465.1 | 2 - 3 | 175 | 74 | |
| XM_012962363.2 | XP_012817817.1 | 2 - 3 | 265 | 74 | ||
| GenBank mRNA | BC166260.1 | - | 2 - 3 | 166 | 74 | |
| BC170606.1 | - | 2 - 3 | 175 | 74 | ||
| BC170985.1 | - | 2 - 3 | 175 | 74 | ||
| CR760937.2 | - | 2 - 3 | 175 | 74 | ||
More Information
Set Membership: |
|

elmod2
Genomic Map